The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_076110.7 | NP_508511.1 | 5 - 6 | 372 | 75 |
GenBank mRNA | AF314195.1 | - | 5 - 6 | 357 | 75 |