Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-26
Entrez Gene ID
180592
Gene Name
TWiK family of potassium channels
Gene Aliases
CELE_C33D12.3
Chromosome Location
Chr.X: 3050365 - 3054824 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076121.7 NP_508522.2 10 - 11 1722 87

View Full Product Details