Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sec-3
Entrez Gene ID
180598
Gene Name
Exocyst complex component 1
Gene Aliases
CELE_F52E4.7
Chromosome Location
Chr.X: 3095428 - 3099852 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076129.6 NP_508530.1 3 - 4 373 66

View Full Product Details