Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- tba-9
- Entrez Gene ID
-
185538
- Gene Name
- Tubulin alpha chain
- Gene Aliases
-
CELE_F40F4.5
- Chromosome Location
-
Chr.X: 3252800 - 3256269 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076154.6 |
NP_508555.2 |
4 - 5 |
583 |
68 |
- Gene Symbol
- lbp-3
- Entrez Gene ID
-
180615
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_F40F4.4
- Chromosome Location
-
Chr.X: 3252889 - 3258155 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383542.1 |
|
4 - 5 |
494 |
68 |
View Full Product Details