Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ddr-2
Entrez Gene ID
180622
Gene Name
protein-coding
Gene Aliases
CELE_F11D5.3
Chromosome Location
Chr.X: 3315319 - 3324242 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076171.6 NP_508572.1 1 - 2 91 107

View Full Product Details