Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wrt-6
Entrez Gene ID
180638
Gene Name
Warthog protein 6 N-product
Gene Aliases
CELE_ZK377.1
Chromosome Location
Chr.X: 3480250 - 3483666 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076196.7 NP_508597.2 1 - 2 140 116

View Full Product Details