Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rgs-7
Entrez Gene ID
180645
Gene Name
protein-coding
Gene Aliases
CELE_F56B6.2
Chromosome Location
Chr.X: 3542542 - 3557783 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029543.5 NP_001024714.1 4 - 5 1061 71
NM_001373260.4 NP_001360776.1 6 - 6 1226 71
NM_001373261.4 NP_001360777.1 5 - 6 1226 71
NM_001373262.3 NP_001360778.1 5 - 5 1137 71
NM_076209.7 NP_508610.3 5 - 6 1226 71
GenBank mRNA AB080201.1 - 4 - 5 1092 71
AY569308.1 - 5 - 6 1137 71

View Full Product Details