Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gst-11
Entrez Gene ID
187817
Gene Name
Glutathione S-Transferase
Gene Aliases
CELE_R11G1.3
Chromosome Location
Chr.X: 3632367 - 3633754 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076224.3 NP_508625.1 3 - 4 344 84

View Full Product Details