Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
abu-10
Entrez Gene ID
185248
Gene Name
Activated in Blocked Unfolded protein response
Gene Aliases
CELE_F35A5.3
Chromosome Location
Chr.X: 3798179 - 3799741 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076258.6 NP_508659.1 2 - 3 433 72

View Full Product Details