Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ttr-48
Entrez Gene ID
191072
Gene Name
TransThyretin-Related family domain
Gene Aliases
CELE_ZC64.2
Chromosome Location
Chr.X: 3840869 - 3842448 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076265.4 NP_508666.1 4 - 5 342 67

View Full Product Details