Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sup-12
Entrez Gene ID
180674
Gene Name
protein-coding
Gene Aliases
CELE_T22B2.4
Chromosome Location
Chr.X: 3904584 - 3907671 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383553.1 3 - 4 423 74
NM_076273.5 NP_508674.1 3 - 4 416 74

View Full Product Details