Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hrg-1
Entrez Gene ID
180684
Gene Name
Heme transporter hrg-1
Gene Aliases
CELE_R02E12.6
Chromosome Location
Chr.X: 4008058 - 4016250 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383555.1 2 - 3 360 106

View Full Product Details