Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clh-4
Entrez Gene ID
180687
Gene Name
protein-coding
Gene Aliases
CELE_T06F4.2
Chromosome Location
Chr.X: 4094867 - 4100573 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029714.2 NP_001024885.1 8 - 9 1536 129
NM_001029715.3 NP_001024886.1 8 - 9 1499 129

View Full Product Details