Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mrp-6
Entrez Gene ID
180691
Gene Name
Uncharacterized protein
Gene Aliases
CELE_F20B6.3
Chromosome Location
Chr.X: 4177839 - 4186756 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001375011.1 NP_001362008.1 10 - 11 2870 92

View Full Product Details