Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hlh-13
Entrez Gene ID
185980
Gene Name
Helix-loop-helix protein 13
Gene Aliases
CELE_F48D6.3
Chromosome Location
Chr.X: 4249249 - 4250766 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076324.5 NP_508725.1 3 - 4 465 59

View Full Product Details