Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mec-18
Entrez Gene ID
180701
Gene Name
Uncharacterized protein
Gene Aliases
CELE_C52B9.9
Chromosome Location
Chr.X: 4271165 - 4274229 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076330.7 NP_508731.3 11 - 12 1520 117

View Full Product Details