Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
amt-4
Entrez Gene ID
180728
Gene Name
Ammonium transporter
Gene Aliases
CELE_C05E11.5
Chromosome Location
Chr.X: 4565302 - 4569620 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076382.6 NP_508783.2 9 - 10 1472 113

View Full Product Details