Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-45
Entrez Gene ID
180738
Gene Name
Nuclear Hormone Receptor family
Gene Aliases
CELE_F16H11.5
Chromosome Location
Chr.X: 4656432 - 4660318 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076396.7 NP_508797.3 7 - 8 1336 111

View Full Product Details