Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acc-3
Entrez Gene ID
186307
Gene Name
Acetylcholine-gated chloride channel subunit acc-3
Gene Aliases
CELE_F55D10.5
Chromosome Location
Chr.X: 4722877 - 4727487 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076409.6 NP_508810.2 6 - 7 735 104
GenBank mRNA AY849796.1 - 6 - 7 749 104

View Full Product Details