Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mls-2
Entrez Gene ID
180751
Gene Name
Homeobox domain-containing protein
Gene Aliases
CELE_C39E6.4
Chromosome Location
Chr.X: 4758804 - 4761640 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076414.7 NP_508815.2 1 - 2 214 144

View Full Product Details