Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mig-13
Entrez Gene ID
180755
Gene Name
protein-coding
Gene Aliases
CELE_F43C9.4
Chromosome Location
Chr.X: 4796087 - 4804118 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029489.6 NP_001024660.1 6 - 7 766 81
NM_001029490.5 NP_001024661.1 4 - 5 652 81
GenBank mRNA AF150958.1 - 5 - 6 670 81

View Full Product Details