Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tsp-14
Entrez Gene ID
180761
Gene Name
protein-coding
Gene Aliases
CELE_F39C12.3
Chromosome Location
Chr.X: 4866681 - 4876965 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380953.1 NP_001367834.1 2 - 3 284 61
NM_001383566.1 2 - 3 312 61

View Full Product Details