Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tyra-3
Entrez Gene ID
187427
Gene Name
protein-coding
Gene Aliases
CELE_M03F4.3
Chromosome Location
Chr.X: 4948049 - 4955358 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029633.5 NP_001024804.1 4 - 5 618 87
NM_001029634.3 NP_001024805.1 4 - 5 757 87
NM_001047810.5 NP_001041275.1 2 - 3 311 87

View Full Product Details