Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hsd-3
Entrez Gene ID
260026
Gene Name
3Beta_HSD domain-containing protein
Gene Aliases
CELE_ZC449.6
Chromosome Location
Chr.X: 5009300 - 5011222 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076451.6 NP_508852.3 3 - 4 651 117

View Full Product Details