Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
zig-6
Entrez Gene ID
192089
Gene Name
Zwei Ig domain protein zig-6
Gene Aliases
CELE_T03G11.8
Chromosome Location
Chr.X: 5186916 - 5189077 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076481.8 NP_508882.3 3 - 4 470 112

View Full Product Details