Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mtm-5
Entrez Gene ID
180792
Gene Name
Myotubularin-related protein 5
Gene Aliases
CELE_H28G03.6
Chromosome Location
Chr.X: 5234669 - 5243408 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076487.4 NP_508888.2 9 - 10 2575 132

View Full Product Details