Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sto-2
Entrez Gene ID
180802
Gene Name
protein-coding
Gene Aliases
CELE_F32A6.5
Chromosome Location
Chr.X: 5307486 - 5323387 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270091.4 NP_001257020.1 5 - 6 658 88
NM_001270092.5 NP_001257021.1 5 - 6 2369 88
NM_001270093.4 NP_001257022.1 5 - 6 651 88
NM_001270094.2 NP_001257023.1 4 - 5 542 88

View Full Product Details