Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ddr-1
Entrez Gene ID
180817
Gene Name
Discoidin domain-containing receptor A
Gene Aliases
CELE_C25F6.4
Chromosome Location
Chr.X: 5461230 - 5466785 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076525.5 NP_508926.2 4 - 5 576 73

View Full Product Details