Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
set-19
Entrez Gene ID
180834
Gene Name
SET domain-containing protein
Gene Aliases
CELE_W01C8.3
Chromosome Location
Chr.X: 5675174 - 5680225 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383587.1 7 - 8 846 78

View Full Product Details