Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-43A1
Entrez Gene ID
180838
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_E03E2.1
Chromosome Location
Chr.X: 5714522 - 5717589 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076560.11 NP_508961.3 3 - 4 491 100

View Full Product Details