Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- puf-9
- Entrez Gene ID
-
180850
- Gene Name
- PUM-HD domain-containing protein
- Gene Aliases
-
CELE_W06B11.2
- Chromosome Location
-
Chr.X: 5841419 - 5845987 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076579.8 |
NP_508980.2 |
8 - 9 |
1939 |
101 |
- Gene Symbol
- atat-2
- Entrez Gene ID
-
180852
- Gene Name
- Alpha-tubulin N-acetyltransferase 2
- Gene Aliases
-
CELE_W06B11.1
- Chromosome Location
-
Chr.X: 5841594 - 5851969 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383590.1 |
|
7 - 8 |
1841 |
101 |
View Full Product Details