Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tbc-16
Entrez Gene ID
180856
Gene Name
Rab-GAP TBC domain-containing protein
Gene Aliases
CELE_F49E10.1
Chromosome Location
Chr.X: 5897338 - 5902157 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076587.7 NP_508988.3 15 - 16 2070 98

View Full Product Details