Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hsp-3
Entrez Gene ID
180880
Gene Name
protein-coding
Gene Aliases
CELE_C15H9.6
Chromosome Location
Chr.X: 6089028 - 6093143 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383595.1 3 - 4 757 59

View Full Product Details