Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lmp-1
Entrez Gene ID
180912
Gene Name
LAMP family protein lmp-1
Gene Aliases
CELE_C03B1.12
Chromosome Location
Chr.X: 6376455 - 6378539 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076671.7 NP_509072.1 4 - 5 617 69
GenBank mRNA AF186848.1 - 3 - 4 456 69

View Full Product Details