Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
igcm-2
Entrez Gene ID
180913
Gene Name
protein-coding
Gene Aliases
CELE_SSSD1.1
Chromosome Location
Chr.X: 6379372 - 6391591 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001375051.2 NP_001362087.1 18 - 19 2253 86
NM_001375052.3 NP_001362088.1 17 - 18 2200 86

View Full Product Details