Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glt-4
Entrez Gene ID
188744
Gene Name
Putative sodium-dependent excitatory amino acid transporter glt-4
Gene Aliases
CELE_T22E5.2
Chromosome Location
Chr.X: 6403424 - 6408411 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076674.7 NP_509075.3 5 - 6 740 119

View Full Product Details