Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ttr-6
Entrez Gene ID
189028
Gene Name
TransThyretin-Related family domain
Gene Aliases
CELE_T28B4.3
Chromosome Location
Chr.X: 6596425 - 6597121 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076709.6 NP_509110.1 1 - 2 83 71

View Full Product Details