Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acr-2
Entrez Gene ID
180936
Gene Name
Acetylcholine receptor subunit beta-type acr-2
Gene Aliases
CELE_K11G12.2
Chromosome Location
Chr.X: 6702549 - 6709308 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076727.9 NP_509128.1 7 - 8 1096 111
GenBank mRNA X86403.1 - - 975 111

View Full Product Details