Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
smf-2
Entrez Gene ID
191766
Gene Name
NRAMP-like transporter smf-2
Gene Aliases
CELE_K11G12.3
Chromosome Location
Chr.X: 6716195 - 6718695 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076730.3 NP_509131.2 3 - 4 479 128

View Full Product Details