Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mdl-1
Entrez Gene ID
180942
Gene Name
BHLH domain-containing protein
Gene Aliases
CELE_R03E9.1
Chromosome Location
Chr.X: 6742351 - 6743899 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076735.6 NP_509136.1 2 - 3 572 107
GenBank mRNA U82968.1 - 2 - 3 437 107

View Full Product Details