Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rig-3
Entrez Gene ID
180958
Gene Name
Neuronal immunoglobulin domain-containing protein rig-3
Gene Aliases
CELE_C53B7.1
Chromosome Location
Chr.X: 6853618 - 6860922 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076754.5 NP_509155.1 8 - 9 1366 96

View Full Product Details