Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-57
Entrez Gene ID
180969
Gene Name
UDPGT domain-containing protein
Gene Aliases
CELE_F01E11.1
Chromosome Location
Chr.X: 6991727 - 6994162 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076781.4 NP_509182.1 6 - 7 1483 152

View Full Product Details