Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tsp-16
Entrez Gene ID
192066
Gene Name
TetraSPanin family
Gene Aliases
CELE_F01E11.4
Chromosome Location
Chr.X: 6994769 - 6996620 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076782.4 NP_509183.2 3 - 4 466 71

View Full Product Details