Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tyra-2
Entrez Gene ID
180970
Gene Name
protein-coding
Gene Aliases
CELE_F01E11.5
Chromosome Location
Chr.X: 6996853 - 7005575 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029351.6 NP_001024522.1 6 - 7 683 91
NM_001038448.5 NP_001033537.1 6 - 7 691 91

View Full Product Details