Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hen-1
Entrez Gene ID
191666
Gene Name
HEsitatioN behavior
Gene Aliases
CELE_C36B7.7
Chromosome Location
Chr.X: 7116180 - 7116996 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076799.6 NP_509200.1 1 - 2 86 80
GenBank mRNA AB067680.1 - 1 - 2 108 80

View Full Product Details