Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pgp-10
Entrez Gene ID
180981
Gene Name
P-GlycoProtein related
Gene Aliases
CELE_C54D1.1
Chromosome Location
Chr.X: 7159036 - 7166021 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076804.5 NP_509205.2 21 - 22 2958 115

View Full Product Details