Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
grd-7
Entrez Gene ID
191661
Gene Name
Ground-like domain-containing protein
Gene Aliases
CELE_F46H5.6
Chromosome Location
Chr.X: 7267870 - 7269479 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076823.5 NP_509224.4 1 - 2 695 60

View Full Product Details