Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spp-3
Entrez Gene ID
180993
Gene Name
Saposin B-type domain-containing protein
Gene Aliases
CELE_T08A9.7
Chromosome Location
Chr.X: 7314965 - 7315353 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076834.7 NP_509235.2 1 - 2 149 71

View Full Product Details