Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spp-4
Entrez Gene ID
188266
Gene Name
Saposin B-type domain-containing protein
Gene Aliases
CELE_T08A9.8
Chromosome Location
Chr.X: 7318239 - 7318798 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076836.5 NP_509237.1 1 - 2 203 84

View Full Product Details