Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sng-1
Entrez Gene ID
180995
Gene Name
Synaptogyrin homolog 1
Gene Aliases
CELE_T08A9.3
Chromosome Location
Chr.X: 7327601 - 7329705 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076838.6 NP_509239.1 3 - 4 345 91
GenBank mRNA AF079373.1 - 3 - 4 338 91

View Full Product Details